Learn More
Strem, An Ascensus Company CAS# 20654-56-2. 1g. Silver perrhenate, 99% (99.995+%-Re) PURATREM. MFCD00014144
SDP
Catalog No. 5090112528
Encompass
CAS# 20654-56-2. 1g. Silver perrhenate, 99% (99.995+%-Re) PURATREM. MFCD00014144. Molecular Weight: 358.07. Molecular Formula: AgReO4. Color/form: white pwdr. Strem# 47-1700. http://www.strem.com/catalog/v/47-1700/
Catalog No. 50-901-12528 Supplier Strem, An Ascensus Company Supplier No. 4717001G
CAS# 20654-56-2. 1g. Silver perrhenate, 99% (99.995+%-Re) PURATREM. MFCD00014144. Molecular Weight: 358.07. Molecular Formula: AgReO4. Color/form: white pwdr. Strem# 47-1700. http://www.strem.com/catalog/v/47-1700/