Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver phosphate, 98%

Catalog No. ac419370250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419370250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC419370250 Supplier Thermo Scientific Chemicals Supplier No. 419370250

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 7784-09-0 | Ag3O4P | 418.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-09-0
Molecular Formula Ag3O4P
Molecular Weight (g/mol) 418.57
MDL Number MFCD00014148
InChI Key FJOLTQXXWSRAIX-UHFFFAOYSA-K
Synonym trisilver phosphate, trisilver 1+ phosphate, unii-zl6t4y1xp8, zl6t4y1xp8, silver orthophosphate, trisilver orthophosphate, silver phosphate ag3po4, silberphosphat, trisilver 1+ orthophosphate, silver orthophosphate ag3po4
PubChem CID 159913
IUPAC Name trisilver;phosphate
SMILES [Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O

Specifications

Melting Point 849°C
Color Green-Yellow to Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 97.5% min
Linear Formula Ag3PO4
Merck Index 15, 8664
Solubility Information Solubility in water: negligible. Other solubilities: slightly sol in dil acetic acid,freely sol in dil nitric acid,in ammonia,,ammonium carbonate,also in alkali cyanides and,thiosulfates
Formula Weight 418.57
Percent Purity 98%
Packaging Glass bottle
Chemical Name or Material Silver phosphate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-049-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.