Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver phosphate, 99% (metals basis)

Catalog No. AA1141506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1141506 5 g
AA1141514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1141506 Supplier Thermo Scientific Chemicals Supplier No. 01141506

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 7784-09-0 | Ag3O4P | 418.57 g/mol

Silver phosphate finds applications in photography, and in silver staining of biological materials. It is used as a photocatalyst for the visible light photochemical splitting of water, and for production of activated oxygen by the same method. It is employed in Crystal growth for scintillation, optics and dosimetry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-09-0
Molecular Formula Ag3O4P
Molecular Weight (g/mol) 418.57
MDL Number MFCD00014148
InChI Key FJOLTQXXWSRAIX-UHFFFAOYSA-K
Synonym trisilver phosphate, trisilver 1+ phosphate, unii-zl6t4y1xp8, zl6t4y1xp8, silver orthophosphate, trisilver orthophosphate, silver phosphate ag3po4, silberphosphat, trisilver 1+ orthophosphate, silver orthophosphate ag3po4
PubChem CID 159913
IUPAC Name trisilver;phosphate
SMILES [Ag+].[Ag+].[Ag+].[O-]P([O-])([O-])=O

Specifications

Melting Point 849°C
Color Gray to Black
Physical Form Powder
Quantity 5 g
Assay Percent Range 99%
Solubility Information Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3,ammonia,ammonium carbonate,alkali cyanides and thiosulfates.
Formula Weight 418.58
Odor Odorless
Sensitivity Light sensitive
Appearance Yellow powder
Density 6.37
Chemical Name or Material Silver phosphate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 232-049-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.