Learn More
Med Vet International Silver-Plated Metal Tracheostomy Tube 3-Piece Kit(Canine), 12mm, J0143D, Each
Catalog No. 5000583286
Encompass_Preferred
German-made. Kit includes:Bulb shaped obturatorInner cannula for easy cleaningOuter cannula with wide outer plate for secure attachment (i.e., umbilical tape)
Catalog No. 50-005-83286 Supplier Med Vet International Supplier No. JOR143D
German-made. Kit includes:Bulb shaped obturatorInner cannula for easy cleaningOuter cannula with wide outer plate for secure attachment (i.e., umbilical tape)