Learn More
Strem, An Ascensus Company CAS# 7440-22-4. 25g. Silver powder (99.95%). MFCD00003397
SDP
Catalog No. 5090115436
Encompass
CAS# 7440-22-4. 25g. Silver powder (99.95%). MFCD00003397. Molecular Weight: 107.87. Molecular Formula: Ag. Color/form: -100 mesh Strem# 93-4755. http://www.strem.com/catalog/v/93-4755/
Catalog No. 50-901-15436 Supplier Strem, An Ascensus Company Supplier No. 93475525G
CAS# 7440-22-4. 25g. Silver powder (99.95%). MFCD00003397. Molecular Weight: 107.87. Molecular Formula: Ag. Color/form: -100 mesh Strem# 93-4755. http://www.strem.com/catalog/v/93-4755/