Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver ShieldR gloves size
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5295591PAK
Encompass
High-tech Norfoil laminate provides a protective barrier from hazardous chemicals. These gloves provide excellent permeation protection against 280 hazardous chemicals and mixtures including acetone benzene epoxy methanol MEK sodium hydroxide sulfuric acid (93%) toluene and trichloroethylene.Gloves feature a five-layer fabric compacted to a thickness of 2.7 mil. Laminating a high-energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35C. Each layer acts as a barrier against various epoxies and industrial chemicals.Lightweight reduces hand fatigue without sacrificing dexterityEconomical - Can be used as a disposable glove without the need for cleaning and re-cyclingDoes not contain latex or chemical accelerators - Little chance of allergic reactionsAmbidextrous hypoallergenicCan be used as a secondary inner glove - Where the dangers of mechanical damage to gloves are
Catalog No. 50-290-1219