Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver, solder alloy, 1.6mm (0.06in) dia

Catalog No. AA4103218
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4103218 50 g
AA4103230 250 g
AA4103236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4103218 Supplier Thermo Scientific Chemicals Supplier No. 04103218

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

For use on copper, stainless, and most other metals

Silver, solder alloy is used on copper, stainless, and most other metals. It is used for electronic assemblies, die attachments, thick films. Soldering of jewellery due to the alloys ability to maintain high levels of brightness.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver, solder Alloy
Name Note 1.6mm (0.06 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Ag:Sn; 3.5:96.5 wt%
Quantity 50 g
MDL Number MFCD00801108
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.