Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver standard solution, for AAS, 1 mg/ml Ag in 0.5N HNO3

Catalog No. AC196301000
Click to view available options
Quantity:
100 mL
500 mL

Catalog No. AC196301000

Supplier: Thermo Scientific Chemicals 196301000

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 7761-88-8 | AgNO3 | 169.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silver standard solution
Name Note 1 mg/ml Ag in 0.5N HNO3
Packaging Plastic bottle
CAS 7761-88-8, 7697-37-2, 7732-18-5
Density 1.0200g/mL
Specific Gravity 1.02
Physical Form Liquid
Molecular Formula AgNO3
Concentration 997 to 1003 μg/mL of Ag (1+) in 0.5N HNO3 (20°C)
MDL Number MFCD00003414
Color Colorless
Melting Point -3.0°C
Merck Index 15, 8657
Boiling Point 101.0°C
Solubility Soluble in water
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [Ag+].[O-][N+]([O-])=O
IUPAC Name silver(1+) nitrate
Molecular Weight (g/mol) 169.87
Quantity 100 mL
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 107.87
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.