Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver sulfadiazine, 98%, Thermo Scientific Chemicals

Catalog No. AC459650050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459650050 5 g
AC459650250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC459650050 Supplier Thermo Scientific Chemicals Supplier No. 459650050
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22199-08-2
Molecular Formula C10H9AgN4O2S
Molecular Weight (g/mol) 357.14
InChI Key UEJSSZHHYBHCEL-UHFFFAOYSA-N
Synonym silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver
PubChem CID 441244
ChEBI CHEBI:9142
IUPAC Name silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide
SMILES C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+]

Specifications

Melting Point 285.0°C
Quantity 5 g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 357.14
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Chemical Name or Material Silver sulfadiazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 244-834-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.