Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Sulfadiazine, Micronized, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18606240 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-240 10 g Amber Glass Bottle
18-606-241 25 g Amber Glass Bottle
18-606-242 100 g Amber Glass Bottle
18-606-243 1 kg Poly Pail
18-606-244 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-240 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S109310GM

S1093, 22199-08-2, C10H9AgN4O2S

Spectrum™ Chemical Silver Sulfadiazine, Micronized, USP is a topical sulfonamide antibacterial used as a topical cream to treat burns. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 22199-08-2
Molecular Formula C10H9AgN4O2S
Molecular Weight (g/mol) 357.14
InChI Key UEJSSZHHYBHCEL-UHFFFAOYSA-N
IUPAC Name silver(1+) 4-{[(pyrimidin-2-yl)azanidyl]sulfonyl}aniline
SMILES [Ag+].NC1=CC=C(C=C1)S(=O)(=O)[N-]C1=NC=CC=N1

Specifications

CAS Max % 1
Quantity 10 g
Percent Purity 98 to 102%
Grade USP
Loss on Drying 0.005
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.