Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Sulfadiazine 98.0+%, TCI America™
SDP

Catalog No. S059525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S059525G 25 g
S05955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S059525G Supplier TCI America Supplier No. S059525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22199-08-2
Molecular Formula C10H9AgN4O2S
Molecular Weight (g/mol) 357.136
MDL Number MFCD00072101
InChI Key UEJSSZHHYBHCEL-UHFFFAOYSA-N
Synonym silver sulfadiazine, sulfadiazine silver, flamazine, silver sulphadiazine, silvadene, sulfadiazine silver salt, dermazin, thermazene, silver sulfadiazinate, sulfadiazine, silver
PubChem CID 441244
ChEBI CHEBI:9142
IUPAC Name silver;(4-aminophenyl)sulfonyl-pyrimidin-2-ylazanide
SMILES C1=CN=C(N=C1)[N-]S(=O)(=O)C2=CC=C(C=C2)N.[Ag+]

Specifications

Melting Point 285°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 357.14
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Silver Sulfadiazine
RTECSNumber WP1950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.