Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Silver sulfate | 10294-26-5 | 99.6% | 311.80 g/mol | Ag2O4S | 100 G
SDP

Catalog No. 5000210121
Encompass_Preferred

Silver sulfate (disilver sulfate) is an analytical-grade inorganic salt supplied for laboratory and biochemical assay applications. It is used in colorimetric determinations and other analytical methods that require a high-purity silver compound.

  • Analytical-grade reagent with high purity.
  • Purity reported as 99.58% (listed as 99.6% rounded).
  • Includes molecular weight of 311.80 g/mol and formula Ag2O4S.
  • Available in multiple pack sizes, including 100 g.
  • Provided with datasheet and certificate of analysis for quality verification.

Catalog No. 50-002-10121 Supplier Medchemexpress LLC Supplier No. HYW051140100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver sulfate (disilver sulfate) is an analytical-grade inorganic salt supplied for laboratory and biochemical assay applications. It is used in colorimetric determinations and other analytical methods that require a high-purity silver compound.

  • Analytical-grade reagent with high purity.
  • Purity reported as 99.58% (listed as 99.6% rounded).
  • Includes molecular weight of 311.80 g/mol and formula Ag2O4S.
  • Available in multiple pack sizes, including 100 g.
  • Provided with datasheet and certificate of analysis for quality verification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.