Learn More
Medchemexpress LLC Silver sulfate | 10294-26-5 | 99.6% | 311.80 g/mol | Ag2O4S | 50 G
SDP
Catalog No. 5000209829
Encompass_Preferred
Silver sulfate (Ag2O4S) is an analytical-grade inorganic reagent used for biochemical assays and analytical determinations. It is typically supplied as a white crystalline powder with high purity and is intended for laboratory use.
  • High purity reagent for analytical applications.
  • White crystalline powder with a defined molecular weight.
  • Suitable for colorimetric determination of nitrates, phosphates, and fluoride.
  • Available in multiple pack sizes, including 50 g.
  • Datasheet and certificate of analysis available.
Catalog No. 50-002-09829 Supplier Medchemexpress LLC Supplier No. HYW05114050G
Silver sulfate (Ag2O4S) is an analytical-grade inorganic reagent used for biochemical assays and analytical determinations. It is typically supplied as a white crystalline powder with high purity and is intended for laboratory use.
  • High purity reagent for analytical applications.
  • White crystalline powder with a defined molecular weight.
  • Suitable for colorimetric determination of nitrates, phosphates, and fluoride.
  • Available in multiple pack sizes, including 50 g.
  • Datasheet and certificate of analysis available.