Learn More
Medchemexpress LLC Silver sulfate | 10294-26-5 | MFCD00003407 | 99.6% | 311.80 g·mol-1 | Ag2O4S | 25 G
SDP
Catalog No. 5000209297
Encompass_Preferred
Silver sulfate is an analytical-grade inorganic reagent used in biochemical and environmental assays, including colorimetric determination of nitrates, phosphates, and fluoride. It is supplied as a high-purity solid with batch documentation available for quality verification, and is intended for research use only.
  • High purity solid (99.6%) suitable for analytical applications.
  • Used for colorimetric determination of nitrates, phosphates, and fluoride.
  • Available in multiple pack sizes including 25 G.
  • Certificate of analysis and safety data sheet available for each batch.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-09297 Supplier Medchemexpress LLC Supplier No. 25GHYW051140
Silver sulfate is an analytical-grade inorganic reagent used in biochemical and environmental assays, including colorimetric determination of nitrates, phosphates, and fluoride. It is supplied as a high-purity solid with batch documentation available for quality verification, and is intended for research use only.
  • High purity solid (99.6%) suitable for analytical applications.
  • Used for colorimetric determination of nitrates, phosphates, and fluoride.
  • Available in multiple pack sizes including 25 G.
  • Certificate of analysis and safety data sheet available for each batch.
  • For research use only; not for human or clinical use.