Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Silver sulfate | 10294-26-5 | MFCD00003407 | 99.6% | 311.80 g/mol | Ag2SO4 | 5 G
SDP

Catalog No. 5000210733
Encompass_Preferred

Silver sulfate is a high-purity inorganic salt used as an analytical reagent for colorimetric determinations and other laboratory assays. It is supplied as a fine, dry powder suitable for research and analytical applications.

  • High purity: 99.6% (typical).
  • Cas number: 10294-26-5.
  • Chemical formula: Ag2SO4; molecular weight: 311.80 g/mol.
  • Pack size: 5 G; solid reagent form.
  • Common applications: colorimetric determination of nitrate, phosphate, and fluoride, and other analytical assays.
  • Storage: store in a cool, dry place, tightly closed.

Catalog No. 50-002-10733 Supplier Medchemexpress LLC Supplier No. HYW0511405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silver sulfate is a high-purity inorganic salt used as an analytical reagent for colorimetric determinations and other laboratory assays. It is supplied as a fine, dry powder suitable for research and analytical applications.

  • High purity: 99.6% (typical).
  • Cas number: 10294-26-5.
  • Chemical formula: Ag2SO4; molecular weight: 311.80 g/mol.
  • Pack size: 5 G; solid reagent form.
  • Common applications: colorimetric determination of nitrate, phosphate, and fluoride, and other analytical assays.
  • Storage: store in a cool, dry place, tightly closed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.