Learn More
Hach Company Silver Sulfate, ACS, 113 g
Supplier:  Hach Company 33414
Encompass
ACS Reagent Grade, CAS No. 10294-26-5, Ag2SO4 Used as a catalyst in Chemical Oxygen Demand determinations
Catalog No. 50-199-4253