Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver Sulfate

Catalog No. 49726610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-726-610G 10 g
49-726-650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 49-726-610G Supplier Sigma Aldrich Supplier No. 49726610G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silver Sulfate

Specifications

Melting Point 652°C
CAS 10294-26-5
Quantity 10 g
Molecular Formula Ag2O4S
Linear Formula Ag2SO4
MDL Number MFCD00003407
Synonym Disilver sulfate; Sulfuric acid disilver(I) salt
Molecular Weight (g/mol) 311.8 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.