Learn More
Silver sulfide, 99%
Description
Silver sulfide is useful as a photosensitizer in photography, Intermediates, chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 825°C |
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99% |
| Solubility Information | Soluble in HNO3,alkali cyanide solutions. Insoluble in water |
| Formula Weight | 247.8 |
| Odor | Odorless |
| Sensitivity | Light sensitive |
| Appearance | Dark gray to black powder/flake/crystalline/beads |
| Show More |
Safety and Handling
| Hazard Category | H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. |
| DOTInformation | DOT Class: 9, Packaging Group: III |
| EINECSNumber | 244-438-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.