Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Tetrafluoroborate, 99%, Spectrum™ Chemical
SDP

Catalog No. 18613229 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-229 5 g Amber Glass Bottle
18-613-228 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-613-229 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S15955GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14104-20-2

Specifications

Color White
Physical Form Crystalline Powder, Needles or Crystals
CAS Max % 1
Quantity 5 g
Odor Odorless
Packaging Amber Glass Bottle
Molecular Formula AgBF4
InChI Key QPRQJCCBNDLHKN-UHFFFAOYSA-N
IUPAC Name Silver(1+) tetrafluoroborate(1-)
Molecular Weight (g/mol) 194.67
Percent Purity 99%
Grade Reagent
Chemical Name or Material Silver Tetrafluoroborate
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.