Learn More
Electron Microscopy Sciences (S-TPPS) Silver Tetraphenylporphin Sulfonate 100 MG
SDP
Catalog No. 5024323
Encompass_Preferred
(S-TPPS)
CAS #31006-50-5 C₄₄H₂₄AgN₄O₁₂S₄⁻⁶
An EM stain for elastin. Albert, E.N., And Fleischer E. (1970). A new electron dense stain for elastic tissue. J. Histochem.Cystochem. 18, 697.
Catalog No. 50-243-23 Supplier Electron Microscopy Sciences Supplier No. 21110
(S-TPPS)
CAS #31006-50-5 C₄₄H₂₄AgN₄O₁₂S₄⁻⁶
An EM stain for elastin. Albert, E.N., And Fleischer E. (1970). A new electron dense stain for elastic tissue. J. Histochem.Cystochem. 18, 697.