Learn More
Strem, An Ascensus Company CAS# 2923-28-6. 5g. Silver trifluoromethanesulfonate, 99% (Silver triflate). MFCD00013226
SDP
Catalog No. 5090112530
Encompass
CAS# 2923-28-6. 5g. Silver trifluoromethanesulfonate, 99% (Silver triflate). MFCD00013226. Molecular Weight: 256.94. Molecular Formula: AgSO3CF3. Color/form: off-white pwdr. Strem# 47-2000. http://www.strem.com/catalog/v/47-2000/
Catalog No. 50-901-12530 Supplier Strem, An Ascensus Company Supplier No. 4720005G
CAS# 2923-28-6. 5g. Silver trifluoromethanesulfonate, 99% (Silver triflate). MFCD00013226. Molecular Weight: 256.94. Molecular Formula: AgSO3CF3. Color/form: off-white pwdr. Strem# 47-2000. http://www.strem.com/catalog/v/47-2000/