Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Silver Trifluoromethanesulfonate

Catalog No. 8532525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-325-25G 5 g
85-325-5G 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 85-325-25G Supplier Sigma Aldrich Supplier No. 8532525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silver trifluoromethanesulfonate p-complexes of monoenes, dienes, trienes, monoynes and diynes have been prepared. It reacts with 2-fluoro- and 3-fluoro-4-alkoxystilbazoles to afford the mesomorphic complexes. Iodine monochloride/AgOTf constitutes an efficient promoter system for the O-glycoside synthesis.Silver trifluoromethanesulfonate (AgOTf) may be employed as a reagent during glucosylation of several alcohols. AgOTf in combination with p-nitrobenzenesulfenyl chloride may be employed as an activator for the glycosylation.It may be used for the synthesis of the following:cystine-containing peptides3-aminoalkylated indolesbenzo[b]oxepines and 2H-chromenesdiversely substituted iminoimidazoazinesReagent for the substitution of halides by triflate.; Reagent for the glycosylation of glycosyl halides; Reagent used for the deprotection of protected thiols

Specifications

CAS 2923-28-6
Molecular Formula CAgF3O3S
Linear Formula CF3SO3Ag
MDL Number MFCD00013226
Quantity 5 g
Synonym Ag(OTf); Silver (trifluoromethyl)sulfonate; Silver triflate; Trifluoromethanesulfonic acid silver salt
Molecular Weight (g/mol) 256.94 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.