Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver tube, 1000mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.95+% | 7440-22-4 | MFCD00003397 | 1EA
Catalog No. 501853130
Encompass
Silver tube, 1000mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 1EA
Catalog No. 50-185-3130 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF791106041EA
Silver tube, 1000mm, outside diameter 5.0mm, inside diameter 4.5mm, wall thickness 0.25mm, as drawn, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 1EA