Learn More
Sigma Aldrich Fine Chemicals Biosciences Silver tube, 100mm, outside diameter 10mm, inside diameter 8mm, wall thickness 1.0mm, as drawn, 99.95+% | 7440-22-4 | MFCD00003397 | 1EA
Catalog No. 501853112
Encompass
Silver tube, 100mm, outside diameter 10mm, inside diameter 8mm, wall thickness 1.0mm, as drawn, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 1EA
Catalog No. 50-185-3112 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF779258191EA
Silver tube, 100mm, outside diameter 10mm, inside diameter 8mm, wall thickness 1.0mm, as drawn, 99.95+% | Purity: 99.95+ % | Mol Wt: 107.87 | 7440-22-4 | MFCD00003397 | 1EA