Learn More
HARVARD BIOSCIENCE INC SILVER WIRE 0.25 MM DIAM
Supplier:  HARVARD BIOSCIENCE INC 641319
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 12.Ag wires made of annealed 99.99% pure silver are supplied for the convenience of researchers to fashion their own electrodes.
Catalog No. NC0937952