Learn More
Strem, An Ascensus Company CAS# 7440-22-4. 1m. Silver wire (99.9%). MFCD00003397
SDP
Catalog No. 5090112505
Encompass
CAS# 7440-22-4. 1m. Silver wire (99.9%). MFCD00003397. Molecular Weight: 107.87. Molecular Formula: Ag. Color/form: 1.0 mm dia. (~8.2g/m) Strem# 47-0250. http://www.strem.com/catalog/v/47-0250/
Catalog No. 50-901-12505 Supplier Strem, An Ascensus Company Supplier No. 4702501MT
CAS# 7440-22-4. 1m. Silver wire (99.9%). MFCD00003397. Molecular Weight: 107.87. Molecular Formula: Ag. Color/form: 1.0 mm dia. (~8.2g/m) Strem# 47-0250. http://www.strem.com/catalog/v/47-0250/