Learn More
World Precision Instrument Teflon Coated Silver Wire-AGT0510
SDP
Catalog No. 50822121
Encompass
Teflon coated silver wire is available is precut lengths. Teflon coating is design to reduce surface friction. Available in 36, 30, and 26-27 AWG.
Catalog No. 50-822-121 Supplier World Precision Instrument Supplier No. AGT0510
Teflon coated silver wire is available is precut lengths. Teflon coating is design to reduce surface friction. Available in 36, 30, and 26-27 AWG.