Learn More
World Precision Instrument Teflon Coated Silver Wire-AGT1510
SDP
Catalog No. 50822127
Encompass
Teflon coated silver wire is available is precut lengths. Teflon coating is design to reduce surface friction. Available in 36, 30, and 26-27 AWG.
Catalog No. 50-822-127 Supplier World Precision Instrument Supplier No. AGT1510
Teflon coated silver wire is available is precut lengths. Teflon coating is design to reduce surface friction. Available in 36, 30, and 26-27 AWG.