Learn More
Costech Analytical Technologies Inc Silvered Cobaltous/cobaltic oxide, 25g
Catalog No. NC9475946
Encompass
Silvered Cobaltous/Cobaltic oxide, 25 gm, Catalysts and Reagent for: NCS and CHNS, Oxygen Analysis, Nitrogen/Protein Analysis, N, N/C and CHN
Catalog No. NC9475946 Supplier Costech Analytical Technologies Inc Supplier No. 011007
Silvered Cobaltous/Cobaltic oxide, 25 gm, Catalysts and Reagent for: NCS and CHNS, Oxygen Analysis, Nitrogen/Protein Analysis, N, N/C and CHN