Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SilverQuest™ Silver Staining Kit

Catalog No. lc6070
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC6070 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC6070 Supplier Invitrogen™ Supplier No. LC6070
Only null left
Add to Cart
Add to Cart

Includes

This kit contains sufficient reagents to stain 25 minigels. Contains: 250 mL Sensitizer, 25 mL Stainer, 250 mL Developer, 2mL Developer Enhancer, 250 mL Stopper solution, 60 mL Destainer A, 60 mL Destainer B

30-fold more sensitive than staining with colloidal Coomassie™ G-250

The SilverQuest™ Silver Staining Kit provides a rapid and easy method to silver stain proteins and DNA in polyacrylamide gels. The SilverQuest™ Silver Staining Kit is specifically designed to provide sensitive silver staining compatible with mass spectrometry analysis.

  • High sensitivity with protein detection at sub-nanogram level
  • Staining compatible with mass spectrometry analysis
  • Rapid and protein bands can be visualized within an hour
  • Easy to use, color-coded solutions for easy staining protocol

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains sufficient reagents to stain 25 minigels

Contains:
250 mL Sensitizer
25 mL Stainer
250 mL Developer
2 mL Developer Enhancer
250 mL Stopper solution
60 mL Destainer A
60 mL Destainer B

Store kit at room temperature (15°C to 30°C). Stable for 6 months.
Detection Location In-Gel Detection
Detection Method Colorimetric
Label or Dye SilverQuest Silver Stain
Quantity 1 kit
Target Molecule Protein
Product Line SilverQuest
Product Type Protein Gel Stain Kit
How much protein is required for mass spectrometry analysis by MALDI-TOF?

In general, the required amount needed to identify a gel-separated protein by enzymatic digestion (usually Trypsin) is about 2 pmole (equivalent to approximately 0.12 µg). Our SilverQuest Silver staining kit is mass spectrometry compatible.

If you need to stop in the middle of the SilverQuest or SilverXpress silver staining procedure, at what point should this be done?

The sensitizing step is the optimal point to stop the procedure. The gel can be left overnight in the Sensitizing Solution. Although some other kits recommend leaving gels in the fix step, we have found that overnight fixation diminishes stain performances.

What is the sensitivity of Coomassie staining and silver staining?

Coomassie sensitivity: 50-500 ng protein per band
Silver-staining sensitivity: 1-5 ng protein per band
In general, silver staining is 10-100 times more sensitive than Coomassie staining.

I used the SilverQuest Silver Staining kit and I see negative staining (a dark halo enclosing a pale area). Can you please help me troubleshoot?

This is usually due to overloading of the protein sample. We recommend decreasing the protein load per band. For an unknown protein, a serial dilution may be necessary to determine the best amount to load for a particular protein.

I used the SilverQuest Silver Staining kit and it took a longer time for band development resulting in a dark background. What happened?

Here are possible causes and solutions:

- Low protein load. Increase the amount of protein load. Be sure to have at least 1-5 ng protein on the gel.
- Some proteins may need longer fixing time. Increase the time for fixing the gel to 2 hours or overnight.

I used the SilverQuest Silver Staining kit and I see a 50-68 kDa band across the gel. What could it be?

This is probably keratin contamination from fingertips or airborne sources. We recommend wearing gloves at all times during electrophoresis and staining steps, and rinsing the gel wells with ultrapure water or running buffer before sample loading.

I used the SilverQuest Silver Staining kit and my stained gels are too dark. What went wrong?

Here are possible causes and solutions:

- Stopper not added to the gel at the appropriate time. Be sure to add the stopper slightly before the desired stain intensity is reached.
- Protein is overloaded. Decrease the protein load on the gel.

I used the SilverQuest Silver Staining kit and see very poor band development or low sensitivity. Can you offer some suggestions?

Here are possible causes and solutions:

- Loss of silver ions from the gel. Limit the wash after staining to 30-60 seconds.
- Stainer or developer solution not prepared properly. Make sure that the solutions are prepared correctly using ultrapure water.
- Low protein load. Increase the amount of protein load. Be sure to have at least 1-5 ng protein on the gel.

I used the SilverQuest Silver Staining kit and obtained a dark/uneven background. What went wrong?

Here are possible causes and solutions:

- Poor water quality. Use ultrapure water of >18 megohm/cm resistance for preparing solutions or rinsing.
- Staining trays not clean or containing solutions left over from prior silver staining. Use staining trays dedicated for silver staining. After silver staining, wash trays with soap and water, and rinse them with ultrapure water.
- Improper washing done between steps. Do not skip or reduce any washing steps.
- Gels are bent or torn. Be careful during handling of the gel. Remove the gels carefully from the cassette after electrophoresis making sure that the gels do not tear.
- Gels are not completely submerged during staining. Be sure to completely immerse gels in staining solution and perform all steps using a rotary shaker for even staining.

How can I decrease the streaking and yellow background that I get with SilverQuest silver staining?

Here are possible causes and solutions:

- High concentration of DTT (>50 mM) in the sample. Use 30-50 mM DTT for reducing the sample.
- To prevent streaking, reduce and alkylate the sample as follows: Reduce the sample with freshly prepared DTT to a final concentration of 17 mM and heat the sample at 70 degrees C for 10 minutes. Then, alkylate the sample with freshly prepared iodoacetamide to a final concentration of 35 mM and heat the sample at 70 degrees C for 10 minutes. Add SDS sample buffer without DTT to the reduced and alkylated sample and proceed with electrophoresis and silver staining.
- Presence of thioflavin in the sensitizer. We recommend heating the 30% ethanol wash in the microwave oven before adding it to the gel after the sensitizing step, and also washing the gel with water a bit longer. You can try this with the turbo method but you risk losing the effects of the sensitizer.

What is the expected level of sensitivity with the SilverQuest Silver Staining Kit?

The kit should be able to detect greater than or equal to 0.3 ng of protein.

Which amino acids are most reactive with the method of silver staining used in SilverXpress and SilverQuest kits?

Glutamic acid, aspartic acid, and cysteine thiols are the most reactive with the SilverXpress and SilverQuest staining methods.

After staining my gel with Colloidal Blue Stain or with SimplyBlue SafeStain, can I then restain my gel with the SilverXpress or Silver Quest staining kit?

Yes this is possible. We recommend destaining the gel with water (it is not necessary to remove all the stain, but if you would like to do so, we recommend soaking the gel in 50% ethanol followed by numerous water washes). If the SimplyBlue stained gel was destained using salt, we recommend washing the gel numerous times in water to remove all the salts before proceeding with the Silver staining protocol. Further, we recommend beginning the silver staining protocol with the fix step (this will help to remove any methanol/ethanol and salts from the previous staining).

Which protein standard do you recommend using with the SilverXpress and SilverQuest Silver Staining kits?

We recommend using the Mark12 Unstained Standard with the SilverXpress and SilverQuest Silver Staining kits. For the SilverQuest Silver Staining kit, we recommend diluting the Mark12 Unstained Standard to 1:10 and loading 5 µL per lane. For the SilverXpress Silver Staining kit, we recommend diluting the Mark12 Unstained Standard to 1:20 and loading 5 µL per lane.

How should I store the SilverXpress and SilverQuest Silver Staining kits and what is their shelf life?

We recommend storing the SilverQuest Silver Staining kit at room temperature and the SilverXpress Silver Staining kit at 4 degrees C. Both kits are stable for one year from date of shipment when properly stored.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.