Learn More
Alkali Scientific SilverSeal CS™ 50 µm Acrylic Adhesive Sealing Film, Sterile, 100/pk
SDP
Catalog No. 502388949
Encompass_Preferred
SilverSeal CS™ 50 µm Acrylic Adhesive Sealing Film are not recommended for PCR or thermocycling. A single non-perforated end tab simplifies application. Simply hold the tab and strip the backing from the body of the sealing film as it lays on the plate to avoid curling. Dimensions are 82.6 by 132.6 mm, including the single 9.5 mm end tab. SilverSeal CS™ sealing films are certified DNase-, RNase-, and nucleic-acid-free. Certified DNase- & RNase- free. Specially formulated for cold storage to -80°C. Excellent barrier properties to retard evaporation. Easily pierceable with pipet tips or robotic probes for sample recovery.
Catalog No. 50-238-8949 Supplier Alkali Scientific Supplier No. SF1SSS
SilverSeal CS™ 50 µm Acrylic Adhesive Sealing Film are not recommended for PCR or thermocycling. A single non-perforated end tab simplifies application. Simply hold the tab and strip the backing from the body of the sealing film as it lays on the plate to avoid curling. Dimensions are 82.6 by 132.6 mm, including the single 9.5 mm end tab. SilverSeal CS™ sealing films are certified DNase-, RNase-, and nucleic-acid-free. Certified DNase- & RNase- free. Specially formulated for cold storage to -80°C. Excellent barrier properties to retard evaporation. Easily pierceable with pipet tips or robotic probes for sample recovery.