Learn More
Alkali Scientific SilverSeal II™ Aluminum Sealing Film, Sterile, 50/pk
SDP
Catalog No. 502389380
Encompass_Preferred
Compared to other aluminum foils, Aluminum Foil Sealing Film has less tendency to roll back on itself when removing the backing paper. Certified DNase- and RNase- free. Excellent chemical resistance (e.g. DMSO). Excellent chemical resistance (e.g. DMSO). 1.4mil conformable aluminum foil allows visual verification of the seal’s security around each well. End-tab removal before thermal cycling is highly recommended. End-tab removal before thermal cycling is highly recommended.
Catalog No. 50-238-9380 Supplier Alkali Scientific Supplier No. SF2SSS
Compared to other aluminum foils, Aluminum Foil Sealing Film has less tendency to roll back on itself when removing the backing paper. Certified DNase- and RNase- free. Excellent chemical resistance (e.g. DMSO). Excellent chemical resistance (e.g. DMSO). 1.4mil conformable aluminum foil allows visual verification of the seal’s security around each well. End-tab removal before thermal cycling is highly recommended. End-tab removal before thermal cycling is highly recommended.