Learn More
Medchemexpress LLC HY-13251 2mg Medchemexpress, Silvestrol CAS:697235-38-4 Purity:>98%
SDP
Catalog No. 502028970
Encompass_Preferred
Medchemexpress, HY-13251 2mg Silvestrol CAS:697235-38-4 Silvestrol is a eukaryotic translation initiation factor 4A (eIF4A) inhibitor isolated from the fruits and twigs of Aglaia foveolata. Silvestrol induces autophagy and caspase-mediated apoptosis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8970 Supplier Medchemexpress LLC Supplier No. HY132512MG
Medchemexpress, HY-13251 2mg Silvestrol CAS:697235-38-4 Silvestrol is a eukaryotic translation initiation factor 4A (eIF4A) inhibitor isolated from the fruits and twigs of Aglaia foveolata. Silvestrol induces autophagy and caspase-mediated apoptosis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.