Learn More
Medchemexpress LLC Silvestrol | 697235-38-4 | >98.0% | 654.66 g/mol | C34H38O13 | 5 MG
SDP
Catalog No. 5000262599
Encompass_Preferred
Silvestrol is a natural eukaryotic translation initiation factor 4A (eIF4A) inhibitor isolated from Agave americana, used as a research reagent to study translation initiation, autophagy, and apoptosis pathways. It exhibits cytotoxic activity in cellular models and is applied in mechanistic studies of protein synthesis and anti-cancer research.
  • Selective eIF4A inhibitor for translation initiation studies.
  • Induces autophagy and caspase-mediated apoptosis in cell models.
  • High purity (>98%) suitable for in vitro experiments.
  • Molecular formula C34H38O13; molecular weight 654.66 g/mol.
  • Supplied in small research quantities for laboratory use.
Catalog No. 50-002-62599 Supplier Medchemexpress LLC Supplier No. HY132515MG
Silvestrol is a natural eukaryotic translation initiation factor 4A (eIF4A) inhibitor isolated from Agave americana, used as a research reagent to study translation initiation, autophagy, and apoptosis pathways. It exhibits cytotoxic activity in cellular models and is applied in mechanistic studies of protein synthesis and anti-cancer research.
  • Selective eIF4A inhibitor for translation initiation studies.
  • Induces autophagy and caspase-mediated apoptosis in cell models.
  • High purity (>98%) suitable for in vitro experiments.
  • Molecular formula C34H38O13; molecular weight 654.66 g/mol.
  • Supplied in small research quantities for laboratory use.