Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Silvestrol | 697235-38-4 | >98.0% | C34H38O13 | 10MG
SDP

Catalog No. 5000261025
Encompass_Preferred

Silvestrol is a natural product isolated from Agave americana that inhibits eukaryotic translation initiation factor 4A (eIF4A) and is used as a research reagent to study translation regulation, autophagy, and apoptosis.

  • Inhibits eIF4A-mediated translation.
  • Induces autophagy and caspase-mediated apoptosis.
  • Exhibits cytotoxic activity in human cancer cell lines.
  • Supplied as a powder for research use.
  • Purity >98.0%.
  • Available in small pack sizes (1 mg-10 mg).
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-61025 Supplier Medchemexpress LLC Supplier No. HY1325110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Silvestrol is a natural product isolated from Agave americana that inhibits eukaryotic translation initiation factor 4A (eIF4A) and is used as a research reagent to study translation regulation, autophagy, and apoptosis.

  • Inhibits eIF4A-mediated translation.
  • Induces autophagy and caspase-mediated apoptosis.
  • Exhibits cytotoxic activity in human cancer cell lines.
  • Supplied as a powder for research use.
  • Purity >98.0%.
  • Available in small pack sizes (1 mg-10 mg).
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.