Learn More
Medchemexpress LLC HY-13250 5mg Medchemexpress, Silvestrol aglycone CAS:960365-65-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132505MG
Encompass_Preferred
Medchemexpress, HY-13250 5mg Silvestrol aglycone CAS:960365-65-5 Silvestrol aglycone is a Silvestrol analogue, inhibits protein translation initiation in cancer cells, with EC50s of 10 and 200 nM for myc-LUC and tub-LUC luciferase reporter protein translation, respectively. Anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1314