Learn More
Bioworld SILWET L-77 10ML
Catalog No. NC1765106
Encompass_Preferred
Silwet L-77 is an organosilicone surfactant. Silwet L-77 is necessary for Agrobacterium-mediated transformation of Aribidopsis and other plants via methods including vacuum-infiltration and floral dip.
Catalog No. NC1765106 Supplier Bioworld Supplier No. 306302161
Silwet L-77 is an organosilicone surfactant. Silwet L-77 is necessary for Agrobacterium-mediated transformation of Aribidopsis and other plants via methods including vacuum-infiltration and floral dip.