Learn More
Bioworld Silwet L-77 | 50mL
Catalog No. NC1791615
Encompass_Preferred
Silwet L-77 is an organosilicone surfactant. Silwet L-77 is necessary for Agrobacterium-mediated transformation of Aribidopsis and other plants via methods including vacuum-infiltration and floral dip mthod.
Catalog No. NC1791615 Supplier Bioworld Supplier No. 306302163
Silwet L-77 is an organosilicone surfactant. Silwet L-77 is necessary for Agrobacterium-mediated transformation of Aribidopsis and other plants via methods including vacuum-infiltration and floral dip mthod.