Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silybin (mixture of Silybin A and Silybin B) 98.0+%, TCI America™
SDP

Catalog No. S050825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S050825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S050825G Supplier TCI America Supplier No. S050825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36804-17-8
Molecular Formula C25H22O10
Molecular Weight (g/mol) 482.441
InChI Key SEBFKMXJBCUCAI-HKTJVKLFSA-N
Synonym Silibinin, 2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-6-(3,5,7-trihydroxy-4-oxobenzopyran-2-yl)benzodioxine
PubChem CID 31553
ChEBI CHEBI:9144
IUPAC Name (2R,3R)-3,5,7-trihydroxy-2-[(2R,3R)-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydro-1,4-benzodioxin-6-yl]-2,3-dihydrochromen-4-one
SMILES COC1=C(C=CC(=C1)C2C(OC3=C(O2)C=C(C=C3)C4C(C(=O)C5=C(C=C(C=C5O4)O)O)O)CO)O

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 25 g
Formula Weight 482.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Silybin (mixture of Silybin A and Silybin B)
RTECSNumber DJ2981770
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.