Learn More
Medchemexpress LLC (±)-Silybin (Silibinin) | 36804-17-8 | 99.4% | 482.44 | 250 MG
SDP
Catalog No. 5000352291
Encompass_Preferred
(±)-Silybin is the racemate of Silybin and is known for inducing apoptosis. It exhibits a range of biological activities including hepatoprotective, antioxidant, anti-inflammatory, and anti-cancer effects. This compound has a chemical formula of C25H22O10, appears as a white to off-white solid, and is classified under Flavonoids and Flavanonols.
  • Induces apoptosis.
  • Exhibits hepatoprotective, antioxidant, anti-inflammatory, and anti-cancer activities.
  • Appears as a white to off-white solid.
  • Classified as a flavonoid and flavanonol.
  • Derived from Leguminosae and Compositae plant families.
Catalog No. 50-003-52291 Supplier Medchemexpress LLC Supplier No. 250MGHY139637
(±)-Silybin is the racemate of Silybin and is known for inducing apoptosis. It exhibits a range of biological activities including hepatoprotective, antioxidant, anti-inflammatory, and anti-cancer effects. This compound has a chemical formula of C25H22O10, appears as a white to off-white solid, and is classified under Flavonoids and Flavanonols.
  • Induces apoptosis.
  • Exhibits hepatoprotective, antioxidant, anti-inflammatory, and anti-cancer activities.
  • Appears as a white to off-white solid.
  • Classified as a flavonoid and flavanonol.
  • Derived from Leguminosae and Compositae plant families.