Learn More
Medchemexpress LLC HY-N0646 1mg Medchemexpress, Silydianin CAS:29782-68-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06461MG
Encompass_Preferred
Medchemexpress, HY-N0646 1mg Silydianin CAS:29782-68-1 Silydianin is an active constituent of Silybium marianum, with exhibit anti-collagenase, antitumor and anti-elastase activities. Silydianin is a natural protein tyrosine phosphatase 1B (PTP1B) with an IC 50 of 17.38 μM. Silydianin has inhibitory effect on the in vitro production and release of oxidative products [1] [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3657