Learn More
Medchemexpress LLC Silymarin | 65666-07-1 | 250 MG
SDP
Catalog No. 5000421988
Encompass_Preferred
Silymarin is an extract derived from milk thistle (Silybum marianum), known for its role as an effective SARS-CoV-2 main protease (Mpro) inhibitor. This compound shows potential in reducing tumor cell proliferation, angiogenesis, and insulin resistance, and exhibits chemopreventive effects on hepatocellular carcinoma (HCC). It is intended for research use only.
  • Acts as an effective SARS-CoV-2 main protease (Mpro) inhibitor
  • Reduces tumor cell proliferation, angiogenesis, and insulin resistance
  • Exhibits chemopreventive effect on hepatocellular carcinoma (HCC)
  • Has potential for COVID-19 research
Catalog No. 50-004-21988 Supplier Medchemexpress LLC Supplier No. 250MGHYN7073
Silymarin is an extract derived from milk thistle (Silybum marianum), known for its role as an effective SARS-CoV-2 main protease (Mpro) inhibitor. This compound shows potential in reducing tumor cell proliferation, angiogenesis, and insulin resistance, and exhibits chemopreventive effects on hepatocellular carcinoma (HCC). It is intended for research use only.
  • Acts as an effective SARS-CoV-2 main protease (Mpro) inhibitor
  • Reduces tumor cell proliferation, angiogenesis, and insulin resistance
  • Exhibits chemopreventive effect on hepatocellular carcinoma (HCC)
  • Has potential for COVID-19 research