Learn More
Bio X Cell InVivoSIM anti-human EGFR, Cetuximab Clone, 5 MG
Supplier: Bio X Cell SIM0002R005MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as Cetuximab, reacting with human EGFR (epidermal growth factor receptor) for research applications. It inhibits tumor cell proliferation by blocking the interaction of EGF with EGFR.
- Human IgG1 isotype
- Purity ≥95% (determined by SDS-PAGE)
- Aggregation <5% (determined by SEC)
- Endotoxin ≤0.5EU/mg
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Protein A purified from cell culture supernatant in an animal-free facility
- Sterilized by 0.2 μm filtration
- Molecular weight 150 kDa
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.