Learn More
Bio X Cell Antibody, Human IgG4, Anti-C5, Eculizumab, 1 MG
Supplier:  Bio X Cell SIM0011R001MG
Encompass_Preferred
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Eculizumab, making it suitable for research. It reacts with human terminal complement component 5 (C5). Eculizumab inhibits the cleavage of C5 to C5α and C5β.
  • Non-therapeutic biosimilar antibody
  • Reacts with human terminal complement component 5 (C5)
  • Inhibits the cleavage of C5
  • Purified from cell culture supernatant in an animal-free facility
  • Sterility confirmed by 0.2 μm filtration
  • Low endotoxin level (≤0.5EU/mg)
  • Low aggregation (<5%)
  • High purity (≥95%)
Catalog No. 50-217-2995