Learn More
Bio X Cell Biosimilar Antibody, human C5 IgG4, 100 MG
Supplier: Bio X Cell SIM0011R100MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as Eculizumab, making it suitable for research applications. It reacts with human terminal complement component 5 (C5), inhibiting its cleavage.
- Clone: Eculizumab
- Unconjugated
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤0.5 EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Isotype: human IgG4
- Purification: Protein A
- Applications: inhibition of C5 activation, functional assays, ELISA, immunoprecipitation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.