Learn More
Bio X Cell InVivoSIM, anti-human C5, Eculizumab, IgG4, 5 MG
Supplier: Bio X Cell SIM0011R005MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Eculizumab, making it suitable for research applications. It specifically targets and reacts with human terminal complement component 5 (C5), inhibiting its cleavage.
- Targets human C5
- Isotype: human IgG4
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤0.5EU/mg
- Purified from cell culture supernatant in an animal-free facility
- Applications include: inhibition of C5 activation, functional assays, ELISA, and immunoprecipitation
- Molecular weight: 150 kDa
- Unconjugated
- Formulation: PBS, pH 7.0, without stabilizers or preservatives
- Sterility: 0.2 μm filtration
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.