Learn More
Bio X Cell Infliximab Anti-TNF Antibody, Human IgG1, 1 MG
Supplier: Bio X Cell SIM0006R001MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Infliximab, making it suitable for research applications. It reacts with human TNFα (tumor necrosis factor-alpha), a multifunctional proinflammatory cytokine involved in various biological processes.
- Reacts with human TNFα
- Isotype: Human IgG1
- Endotoxin: ≤0.5 EU/mg
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze
- Reported applications: in vitro TNFα neutralization, in vitro functional assay, Flow Cytometry, ELISA, Western Blot
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.