Learn More
Bio X Cell InVivoSIM anti-human TNF (Infliximab Biosimilar), Human IgG1, 95% Purity, 5MG, 5 MG
Supplier: Bio X Cell SIM0006R005MG
This non-therapeutic biosimilar antibody utilizes the same variable regions as the therapeutic antibody Infliximab, making it suitable for research applications. It reacts with human TNF, a multifunctional proinflammatory cytokine. This biosimilar blocks the interaction of TNF with its receptors TNFR1 (p55) and TNFR2 (p75), down-regulating the inflammatory response associated with autoimmune diseases.
- Human IgG1 isotype
- Reacts w/ human TNF
- Immunogen: human TNF
- Endotoxin level 0.5 EU/mg
- Purity 95%
- Sterility via 0.2 m filtration
- Purified from cell culture supernatant in an animal-free facility
- Purification method: Protein A
- Formulated in PBS, pH 7.0, w/o stabilizers or preservatives
- Mol weight: 150 kDa
- Reported applications include in vitro TNF neutralization, in vitro functional assay, Flow Cytometry, ELISA, and Western Blot
- Store at stock concentration at 4C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.