Learn More
NEOGEN SIM Medium, 500 g
SDP
Catalog No. 502015921
Encompass_Preferred
SIM Medium is used for the differentiation of microorganisms on the basis of hydrogen sulfide production, indole production, and motility in a laboratory setting. SIM Medium is not intended for use in the diagnosis of disease or other conditions in humans. Semisolid media have been used extensively in the determination of bacterial motility. The production of hydrogen sulfide, indole formation, and motility are useful diagnostic tests in the identification of Enterobacteriaceae, especially Salmonella spp. and Shigella spp. In 1940, Sulkin and Willett demonstrated motility, hydrogen sulfide production, and carbohydrate fermentation by members of Salmonella and Shigella groups. They called attention to the “brush-like growth” or motility of the typhoid organisms. Greene et al. used SIM Medium to detect motility in a large series of cultures of typhoid organisms.For more information visit https://www.neogen.com/solutions/Microbiology/sim-medium
Catalog No. 50-201-5921 Supplier NEOGEN Supplier No. 700003636
Add to Cart
Add to Cart
SIM Medium is used for the differentiation of microorganisms on the basis of hydrogen sulfide production, indole production, and motility in a laboratory setting. SIM Medium is not intended for use in the diagnosis of disease or other conditions in humans. Semisolid media have been used extensively in the determination of bacterial motility. The production of hydrogen sulfide, indole formation, and motility are useful diagnostic tests in the identification of Enterobacteriaceae, especially Salmonella spp. and Shigella spp. In 1940, Sulkin and Willett demonstrated motility, hydrogen sulfide production, and carbohydrate fermentation by members of Salmonella and Shigella groups. They called attention to the “brush-like growth” or motility of the typhoid organisms. Greene et al. used SIM Medium to detect motility in a large series of cultures of typhoid organisms.For more information visit https://www.neogen.com/solutions/Microbiology/sim-medium