Learn More
Alkali Scientific SIM Medium 8mL Fill 16x125mm Screw Cap, 10/Pk
SDP
Catalog No. 5000663130
Encompass_Preferred
SIM Medium, 8mL Fill, 16x125mm Screw Cap Tube, 10/Pk is a multipurpose differential medium used to evaluate sulfide production, indole formation, and motility in microorganisms, particularly members of the Enterobacteriaceae family. The formulation supports bacterial growth while enabling several key biochemical reactions used in microbial identification workflows. Black precipitate formation indicates hydrogen sulfide production, while indole formation can be detected after reagent addition. Motility is observed by the diffusion of growth away from the stab line in the medium. Each tube contains 8mL of prepared medium in a 16x125mm screw cap tube suitable for incubation and routine laboratory handling. This ready to use format supports microbial identification procedures in clinical, research, and quality control laboratories.
Catalog No. 50-006-63130 Supplier Alkali Scientific Supplier No. SIMT00
SIM Medium, 8mL Fill, 16x125mm Screw Cap Tube, 10/Pk is a multipurpose differential medium used to evaluate sulfide production, indole formation, and motility in microorganisms, particularly members of the Enterobacteriaceae family. The formulation supports bacterial growth while enabling several key biochemical reactions used in microbial identification workflows. Black precipitate formation indicates hydrogen sulfide production, while indole formation can be detected after reagent addition. Motility is observed by the diffusion of growth away from the stab line in the medium. Each tube contains 8mL of prepared medium in a 16x125mm screw cap tube suitable for incubation and routine laboratory handling. This ready to use format supports microbial identification procedures in clinical, research, and quality control laboratories.